accessibility menu, dialog, popup

UserName

CAS RN 98-07-7

IUPAC Name: (trichloromethyl)benzene
Synonyms: benzotrichloride
Molecular Weight (g/mol): 195.47
Molecular Formula: C7H5Cl3
InChi Key: XEMRAKSQROQPBR-UHFFFAOYSA-N
SMILES: ClC(Cl)(Cl)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)