accessibility menu, dialog, popup

UserName

CAS RN 97840-40-9

IUPAC Name: 4-(3-chlorophenoxy)piperidine
Synonyms: 4-(3-chlorophenoxy)piperidine
Molecular Weight (g/mol): 211.69
Molecular Formula: C11H14ClNO
InChi Key: BYXWGMQMUJCYPK-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(OC2CCNCC2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results