accessibility menu, dialog, popup

UserName

CH3OClOH3C

CAS RN 97-97-2

IUPAC Name: 2-chloro-1,1-dimethoxyethane
Synonyms: 2-chloro-1,1-dimethoxyethane
Molecular Weight (g/mol): 124.56
Molecular Formula: C4H9ClO2
InChi Key: CRZJPEIBPQWDGJ-UHFFFAOYSA-N
SMILES: COC(CCl)OC
Boiling Point 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)