missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Chloroacetal 98.0+%, TCI America™

Catalog No. C0085500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0085500ML 500 mL
C008525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0085500ML Supplier TCI America Supplier No. C0085500ML
Only null left

Chemical Identifiers

CAS 97-97-2
Molecular Formula C4H9ClO2
Molecular Weight (g/mol) 124.56
MDL Number MFCD00000948
InChI Key CRZJPEIBPQWDGJ-UHFFFAOYSA-N
Synonym chloroacetaldehyde dimethyl acetal, dimethylchloroacetal, dimethyl chloroacetal, dimethyl chloracetal, ethane, 2-chloro-1,1-dimethoxy, 2-chloro-1,1-dimethoxy-ethane, chloroacetaldehyde, dimethyl acetal, 2-chloroacetaldehyde, dimethyl acetal, methyl chloroacetal, 1-chloro-2,2-dimethoxyethane
PubChem CID 60986
IUPAC Name 2-chloro-1,1-dimethoxyethane
SMILES COC(CCl)OC

Specifications

Color Yellow
Boiling Point 130°C
Quantity 500 mL
UN Number 1993
Formula Weight 124.56
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Chloroacetal
EINECSNumber (2)-3589
TSCA Yes