accessibility menu, dialog, popup

UserName

H3COOCH3CH3

CAS RN 97-62-1

IUPAC Name: ethyl 2-methylpropanoate
Synonyms: ethyl isobutyrate
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: WDAXFOBOLVPGLV-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C)C
Percent Purity 
  • (4)
  • (3)
  • (2)
Boiling Point 
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (4)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (2)