missing translation for 'onlineSavingsMsg'
Learn More

Ethyl isobutyrate, 98%

Catalog No. AAA12966AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA12966AP 500 mL
AAA12966AE 100 mL
AAA129660F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA12966AP Supplier Thermo Scientific Chemicals Supplier No. A12966AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 97-62-1 | C6H12O2 | 116.16 g/mol

It is employed to make flavoring compounds and essences, as a flavoring agent and to flavor animal feeds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed to make flavoring compounds and essences, as a flavoring agent and to flavor animal feeds.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from ignition sources. Keep away from electrostatic charges.

Chemical Identifiers

CAS 97-62-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009165
InChI Key WDAXFOBOLVPGLV-UHFFFAOYSA-N
Synonym ethyl isobutyrate, ethyl isobutanoate, ethylisobutyrate, isobutyric acid ethyl ester, ethyl 2,2-dimethylacetate, propanoic acid, 2-methyl-, ethyl ester, isobutyric acid, ethyl ester, ethyl 2-methylpropionate, propionic acid, 2-methyl-, ethyl ester, ethyl isobutyrate natural
PubChem CID 7342
ChEBI CHEBI:87303
IUPAC Name ethyl 2-methylpropanoate
SMILES CCOC(=O)C(C)C

Specifications

Melting Point -88°C
Density 0.869
Boiling Point 108°C to 110°C
Flash Point 13°C (55°F)
Odor Pleasant
Refractive Index 1.387
Quantity 500 mL
UN Number UN2385
Beilstein 773846
Merck Index 14,3814
Solubility Information Not miscible or difficult to mix in water. Soluble in alcohol.
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material Ethyl isobutyrate
Show More Show Less
Hazard Category H225-H315-H319
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYL ISOBUTYRATE
EINECSNumber 202-595-4
RTECSNumber NQ4675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only