accessibility menu, dialog, popup

UserName

CAS RN 97-61-0

IUPAC Name: 2-methylpentanoic acid
Synonyms: pentanoic acid, 2-methyl-
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: OVBFMEVBMNZIBR-UHFFFAOYNA-N
SMILES: CCCC(C)C(O)=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)