missing translation for 'onlineSavingsMsg'
Learn More

2-Acide méthylvalérique 98,0+%, TCI America™

Catalog No. M0610500ML
Change view
Click to view available options
Quantité:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
M0610500ML 500 mL
M061025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0610500ML Supplier TCI America Supplier No. M0610500ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 97-61-0
Formule moléculaire C6H12O2
Poids moléculaire (g/mol) 116.16
Numéro MDL MFCD00002671
Clé InChI OVBFMEVBMNZIBR-UHFFFAOYSA-N
Synonyme 2-methylvaleric acid, pentanoic acid, 2-methyl, valeric acid, 2-methyl, methylpropylacetic acid, 2-pentanecarboxylic acid, 2-methyl valeric acid, alpha-methylvaleric acid, pentanoic acid, methyl, kyselina 2-methylvalerova, fema no. 2754
PubChem CID 7341
Nom de l’IUPAC Acide 2-méthylpentanoïque
SOURIRES CCCC(C)C(=O)O

Specifications

Nom ou substance chimique 2-Methylvaleric Acid
Couleur Jaune
Point d’ébullition 195 °C
Quantité 500 mL
Numéro UN 3265
Poids de formule 116.16
Pourcentage de pureté ≥98,0% (GC,T)
Forme physique Liquide
EINECSNumber (2)-0608
RTECSNumber YV7700000
TSCA Oui