accessibility menu, dialog, popup

UserName

SONON

CAS RN 96989-50-3

IUPAC Name: bis(pyridin-2-yloxy)methanethione
Synonyms: bis(pyridin-2-yloxy)methanethione
Molecular Weight (g/mol): 232.26
Molecular Formula: C11H8N2O2S
InChi Key: IKYOVSVBLHGFMA-UHFFFAOYSA-N
SMILES: S=C(OC1=CC=CC=N1)OC1=CC=CC=N1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (5)
  • (4)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)