missing translation for 'onlineSavingsMsg'
Learn More

O,O'-Di-2-pyridyl Thiocarbonate 98.0+%, TCI America™

Catalog No. T19061G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T19061G Supplier TCI America Supplier No. T19061G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 96989-50-3
Molecular Formula C11H8N2O2S
Molecular Weight (g/mol) 232.257
MDL Number MFCD00074870
InChI Key IKYOVSVBLHGFMA-UHFFFAOYSA-N
Synonym di-2-pyridyl thionocarbonate, o,o-di pyridin-2-yl carbonothioate, o,o'-di-2-pyridyl thiocarbonate, o,o-dipyridin-2-yl carbonothioate, di 2-pyridyl thionocarbonate, carbonothioic acid, o,o-di-2-pyridinyl ester, di 2-pyridyloxy methane-1-thione, o,o-dipyridin-2-yl thiocarbonate, bis pyridin-2-yloxy methanethione, di-2-pyridyl thiocarbomate
PubChem CID 719784
IUPAC Name dipyridin-2-yloxymethanethione
SMILES C1=CC=NC(=C1)OC(=S)OC2=CC=CC=N2

Specifications

Melting Point 99°C
Quantity 1 g
Formula Weight 232.26
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material O,O′-Di-2-pyridyl Thiocarbonate
TSCA No
Recommended Storage Refrigerator