accessibility menu, dialog, popup

UserName

CAS RN 96859-34-6

IUPAC Name: 3-(2H-1,2,3,4-tetrazol-5-yl)phenol
Synonyms: 3-(2H-1,2,3,4-tetrazol-5-yl)phenol
Molecular Weight (g/mol): 162.15
Molecular Formula: C7H6N4O
InChi Key: IZORRBUQWFSCII-UHFFFAOYSA-N
SMILES: OC1=CC=CC(=C1)C1=NNN=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results