missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Tetrazol-5-yl)phenol, 97%

Catalog No. AAH27529MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27529MD 250 mg
AAH2752903 1 g
AAH2752906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH27529MD Supplier Thermo Scientific Chemicals Supplier No. H27529MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 96859-34-6 | C7H6N4O | 162.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 96859-34-6
Molecular Formula C7H6N4O
Molecular Weight (g/mol) 162.15
MDL Number MFCD06797247
InChI Key IZORRBUQWFSCII-UHFFFAOYSA-N
Synonym 3-1h-tetrazol-5-yl phenol, 5-3-hydroxyphenyl tetrazole, 3-2h-tetrazol-5-yl phenol, 3-2h-1,2,3,4-tetrazol-5-yl phenol, 3-1h-1,2,3,4-tetrazol-5-yl phenol, m-tetrazolylphenol, 3-2h-tetrazol-5-yl-phenol, 5-3-hydroxyphenyl-1h-tetrazole
PubChem CID 13455763
SMILES OC1=CC=CC(=C1)C1=NNN=N1

Specifications

Melting Point ∽220°C (decomposition)
Quantity 250 mg
IUPAC Name 3-(2H-tetrazol-5-yl)phenol
Formula Weight 162.16
Percent Purity 97%
Chemical Name or Material 3-(1H-Tetrazol-5-yl)phenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only