accessibility menu, dialog, popup

UserName

OHHOHS

CAS RN 96-27-5

IUPAC Name: 3-sulfanylpropane-1,2-diol
Synonyms: thioglycerol 3-mercapto-1,2-propanediole thiovanol thioglycerine monothioglycerin thioglycerin usaf B-40 usaf CB-37
Molecular Weight (g/mol): 108.16
Molecular Formula: C3H8O2S
InChi Key: PJUIMOJAAPLTRJ-UHFFFAOYNA-N
SMILES: OCC(O)CS
Percent Purity 
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (7)
  • (3)
  • (3)
Boiling Point 
  • (2)
  • (3)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (1)
  • (3)
  • (6)
  • (5)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (7)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)