missing translation for 'onlineSavingsMsg'
Learn More

alpha-Thioglycerol 95.0+%, TCI America™

Catalog No. T0905500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0905500G 500 g
T090525G 25 g
T0905100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0905500G Supplier TCI America Supplier No. T0905500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 96-27-5
Molecular Formula C3H8O2S
Molecular Weight (g/mol) 108.16
MDL Number MFCD00004879
InChI Key PJUIMOJAAPLTRJ-UHFFFAOYNA-N
Synonym 3-mercapto-1,2-propanediol, monothioglycerol, 1-thioglycerol, thioglycerol, thioglycerine, thioglycerin, thiovanol, 3-mercaptopropane-1,2-diol, alpha-thioglycerol, glycerol-1-thiol
PubChem CID 7291
ChEBI CHEBI:74537
IUPAC Name 3-sulfanylpropane-1,2-diol
SMILES OCC(O)CS

Specifications

Color Colorless
Boiling Point 139°C
Quantity 500 g
UN Number 2810
Formula Weight 108.16
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material alpha-Thioglycerol
EINECSNumber (9)-0805
RTECSNumber TY8140000
TSCA Yes