missing translation for 'onlineSavingsMsg'
Learn More

alpha-Monothioglycerol, 99.1%, For GC analysis, MP Biomedicals™

Catalog No. ICN15572325
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15572325 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15572325 Supplier MP Biomedicals Inc Supplier No. 0215572325

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 96-27-5
Molecular Formula C3H8O2S
Molecular Weight (g/mol) 108.16
MDL Number MFCD00004879
InChI Key PJUIMOJAAPLTRJ-UHFFFAOYNA-N
Synonym 3-mercapto-1,2-propanediol, monothioglycerol, 1-thioglycerol, thioglycerol, thioglycerine, thioglycerin, thiovanol, 3-mercaptopropane-1,2-diol, alpha-thioglycerol, glycerol-1-thiol
PubChem CID 7291
ChEBI CHEBI:74537
IUPAC Name 3-sulfanylpropane-1,2-diol
SMILES OCC(O)CS

Specifications

Color Colorless
Quantity 25 mL
Formula Weight 108.2
Percent Purity 0.991
Physical Form Liquid
Chemical Name or Material α-Monothioglycerol, For GC analysis