missing translation for 'onlineSavingsMsg'
Learn More

Monothioglycerol, NF, 97-101%, Spectrum™ Chemical

Catalog No. 18604693
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604693 500 mL Amber Glass Bottle
18604691 25 mL Amber Glass Bottle
18604692 100 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18604693 Supplier Spectrum Chemical Mfg Cor Supplier No. M4080500ML
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

M4080, 96-27-5, C3H8O2S

Spectrum™ Chemical Monothioglycerol, NF

Chemical Identifiers

CAS 96-27-5
Molecular Formula C3H8O2S
Molecular Weight (g/mol) 108.16
MDL Number MFCD00004879
InChI Key PJUIMOJAAPLTRJ-UHFFFAOYNA-N
IUPAC Name 3-sulfanylpropane-1,2-diol
SMILES OCC(O)CS

Specifications

CAS Max % 1
pH 3.5 to 7.0
Boiling Point 118°C (244.2°F)
Ignition Residue 0.001
Packaging Amber Glass Bottle
Refractive Index 1.521-1.526
Quantity 500 mL
Specific Gravity 1.241-1.250
Percent Purity 97 to 101%
Grade NF
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Monothioglycerol
Show More Show Less