missing translation for 'onlineSavingsMsg'
Learn More

3-Mercapto-1,2-propanediol, 90%

Catalog No. AAA1894830
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1894830 250 g
AAA1894818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1894830 Supplier Thermo Scientific Chemicals Supplier No. A1894830
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 96-27-5 | C3H8O2S | 108.16 g/mol

It generally used as a size-regulating capping agent for nanocrystals. 1-Thioglycerol is also used in bacteriology experiments to induce the synthesis of porphyrin compounds in aerobically growing Escherichia coli. Also used to study the pH-sensitive photoluminescence of aqueous thiol-capped CdTe nanocrystals. 1-Thioglycerol is used to develop and test thiol functionalized copolymers. 1-Thioglycerol is used as a post-modification agent in the generation of non-standard peptide foldamers.Potential substitute for 2-mercaptoethanol, probe for the study of lymphocyte activation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It generally used as a size-regulating capping agent for nanocrystals. 1-Thioglycerol is also used in bacteriology experiments to induce the synthesis of porphyrin compounds in aerobically growing Escherichia coli. Also used to study the pH-sensitive photoluminescence of aqueous thiol-capped CdTe nanocrystals. 1-Thioglycerol is used to develop and test thiol functionalized copolymers. 1-Thioglycerol is used as a post-modification agent in the generation of non-standard peptide foldamers.Potential substitute for 2-mercaptoethanol, probe for the study of lymphocyte activation.

Solubility
Slightly soluble in water and benzene. Soluble in acetone. Insoluble in ether.

Notes
Air Sensitive and Hygroscopic, store away from water/moisture and air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.

Chemical Identifiers

CAS 96-27-5
Molecular Formula C3H8O2S
Molecular Weight (g/mol) 108.16
MDL Number MFCD00004879
InChI Key PJUIMOJAAPLTRJ-UHFFFAOYNA-N
Synonym 3-mercapto-1,2-propanediol, monothioglycerol, 1-thioglycerol, thioglycerol, thioglycerine, thioglycerin, thiovanol, 3-mercaptopropane-1,2-diol, alpha-thioglycerol, glycerol-1-thiol
PubChem CID 7291
ChEBI CHEBI:74537
IUPAC Name 3-sulfanylpropane-1,2-diol
SMILES OCC(O)CS

Specifications

Density 1.247
Boiling Point 117°C to 119°C (5 mmHg)
Flash Point 149°C (300°F)
Refractive Index 1.511
Linear Formula HOCH2CH(OH)CH2SH
Quantity 250 g
Beilstein 1732046
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,9335
Solubility Information Slightly soluble in water and benzene. Soluble in acetone. Insoluble in ether.
Formula Weight 108.16
Percent Purity ≥90%
Chemical Name or Material 3-Mercapto-1,2-propanediol
Show More Show Less
Hazard Category H302-H311-H315-H317-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 202-495-0
RTECSNumber TY8140000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only