accessibility menu, dialog, popup

UserName

CAS RN 96-23-1

IUPAC Name: 1,3-dichloropropan-2-ol
Synonyms: 1,3-dichloro-2-propanol glycerol dichlorohydrin glycerol 1,3-dichlorohydrin dichlorohydrin 1,3-dichlorohydrin C3H6cl2O α,gamma-dichlorohydrin enodrin sym-dichloroisopropyl alcohol Gdch
Molecular Weight (g/mol): 128.98
Molecular Formula: C3H6Cl2O
InChi Key: DEWLEGDTCGBNGU-UHFFFAOYSA-N
SMILES: OC(CCl)CCl
Boiling Point 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)