missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-2-propanol, 98+%

Catalog No. AAA1600330
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1600330 250 g
AAA160030B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1600330 Supplier Thermo Scientific Chemicals Supplier No. A1600330
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 96-23-1 | C3H6Cl2O | 128.98 g/mol

1,3-Dichloro-2-propanol biotransformation to epichlorohydrin by the whole cells of recombinant Escherichia coli via resin-based in situ product removal has been investigated. It is used in the synthesis of glycerol, in the production of plastics and textiles and in the synthesis of pharmaceuticals. Also used as a solvent and as a cement for celluloid. It is also present in some foods, e.g. soy sauce, soup spices and instant soups, from hydrolization of proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dichloro-2-propanol biotransformation to epichlorohydrin by the whole cells of recombinant Escherichia coli via resin-based in situ product removal has been investigated. It is used in the synthesis of glycerol, in the production of plastics and textiles and in the synthesis of pharmaceuticals. Also used as a solvent and as a cement for celluloid. It is also present in some foods, e.g. soy sauce, soup spices and instant soups, from hydrolization of proteins.

Solubility
Fully miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 96-23-1
Molecular Formula C3H6Cl2O
Molecular Weight (g/mol) 128.98
MDL Number MFCD00000951
InChI Key DEWLEGDTCGBNGU-UHFFFAOYSA-N
Synonym 1,3-dichloro-2-propanol, dichlorohydrin, 1,3-dichloroisopropanol, 1,3-dichlorohydrin, glycerol 1,3-dichlorohydrin, 2-propanol, 1,3-dichloro, enodrin, 1,3-dichloro-2-hydroxypropane, 1,3-dichloroisopropyl alcohol, sym-glycerol dichlorohydrin
PubChem CID 7289
ChEBI CHEBI:18917
IUPAC Name 1,3-dichloropropan-2-ol
SMILES OC(CCl)CCl

Specifications

Melting Point -4°C
Density 1.366
Boiling Point 174°C to 176°C
Flash Point 85°C (185°F)
Refractive Index 1.483
Linear Formula ClCH2CH(OH)CH2Cl
Quantity 250 g
UN Number UN2750
Beilstein 1732063
Merck Index 14,3074
Solubility Information Fully miscible in water.
Formula Weight 128.99
Percent Purity ≥98%
Chemical Name or Material 1,3-Dichloro-2-propanol
Show More Show Less
Hazard Category H227-H301-H312-H350
Hazard Statement GHS H Statement
H301-H311-H350-H227
Toxic if swallowed.
Toxic in contact with skin.
May cause cancer.
Combustible liquid.
Precautionary Statement P201-P202-P210-P264b-P270-P281-P301+P310-P302+P352-P308+P313-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 1,3-DICHLOROPROPANOL-2
EINECSNumber 202-491-9
RTECSNumber UB1400000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only