accessibility menu, dialog, popup

UserName

HOBrBr

CAS RN 96-21-9

IUPAC Name: 1,3-dibromopropan-2-ol
Synonyms: 1,3-dibromo-2-propanol
Molecular Weight (g/mol): 217.89
Molecular Formula: C3H6Br2O
InChi Key: KIHQZLPHVZKELA-UHFFFAOYSA-N
SMILES: OC(CBr)CBr
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)