accessibility menu, dialog, popup

UserName

BOOCH3CH3CH3CH3NCH3CH3BOOH3CH3CH3CH3C

CAS RN 958261-51-3

IUPAC Name: 9-(heptadecan-9-yl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
Synonyms: 9-(heptadecan-9-yl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
Molecular Weight (g/mol): 657.59
Molecular Formula: C41H65B2NO4
InChi Key: XMKFCPVHTTWWCK-UHFFFAOYSA-N
SMILES: CCCCCCCCC(CCCCCCCC)N1C2=C(C=CC(=C2)B2OC(C)(C)C(C)(C)O2)C2=C1C=C(C=C2)B1OC(C)(C)C(C)(C)O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results