missing translation for 'onlineSavingsMsg'
Learn More

9-(9-Heptadecanyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole 97.0+%, TCI America™

Catalog No. O0428200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0428200MG 200 mg
O04281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O0428200MG Supplier TCI America Supplier No. O0428200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 958261-51-3
Molecular Formula C41H65B2NO4
Molecular Weight (g/mol) 657.594
MDL Number MFCD16621134
InChI Key XMKFCPVHTTWWCK-UHFFFAOYSA-N
Synonym 9-(1-n-Octylnonyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole, 9-(1-n-Octylnonyl)carbazole-2,7-diboronic Acid Bis(pinacol) Ester, 9-(9-Heptadecanyl)carbazole-2,7-diboronic Acid Bis(pinacol) Ester
PubChem CID 51341870
IUPAC Name 9-heptadecan-9-yl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=C(N3C(CCCCCCCC)CCCCCCCC)C=C(C=C4)B5OC(C(O5)(C)C)(C)C

Specifications

Melting Point 146°C
Color White
Quantity 200 mg
Formula Weight 657.59
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 9-(9-Heptadecanyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole