accessibility menu, dialog, popup

UserName

CAS RN 95713-52-3

IUPAC Name: (2S)-2-[(5-fluoro-2,4-dinitrophenyl)amino]propanamide
Synonyms: (2S)-2-[(5-fluoro-2,4-dinitrophenyl)amino]propanamide
Molecular Weight (g/mol): 272.19
Molecular Formula: C9H9FN4O5
InChi Key: NEPLBHLFDJOJGP-BYPYZUCNSA-N
SMILES: C[C@H](NC1=CC(F)=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(N)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results