missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-(5-Fluoro-2,4-dinitrophenyl)-L-alaninamide 95.0+%, TCI America™

Catalog No. D22591G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22591G 1 g
D2259100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D22591G Supplier TCI America Supplier No. D22591G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 95713-52-3
Molecular Formula C9H9FN4O5
Molecular Weight (g/mol) 272.19
MDL Number MFCD00042049
InChI Key NEPLBHLFDJOJGP-BYPYZUCNSA-N
Synonym marfey's reagent, s-2-5-fluoro-2,4-dinitrophenyl amino propanamide, 2s-2-5-fluoro-2,4-dinitrophenyl amino propanamide, n-2,4-dinitro-5-fluorophenyl-l-alaninamide, 2s-2-5-fluoro-2,4-dinitroanilino propanamide, propanamide, 2-5-fluoro-2,4-dinitrophenyl amino-, 2s, nalpha-2,4-dinitro-5-fluorophenyl-l-alaninamide, n-alpha-2,4-dinitro-5-fluorophenyl-l-alaninamide, fdaa, 1-fd-aa
PubChem CID 5486955
IUPAC Name (2S)-2-[(5-fluoro-2,4-dinitrophenyl)amino]propanamide
SMILES C[C@H](NC1=CC(F)=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(N)=O

Specifications

Melting Point 229°C
Color Yellow
Quantity 1 g
Formula Weight 272.19
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-(5-Fluoro-2,4-dinitrophenyl)-L-alaninamide [HPLC Labeling Reagent for e.e. Determination]