accessibility menu, dialog, popup

UserName

ClO(E)

CAS RN 956-02-5

IUPAC Name: (2E)-1-(4-chlorophenyl)-3-phenylprop-2-en-1-one
Synonyms: (2E)-1-(4-chlorophenyl)-3-phenylprop-2-en-1-one
Molecular Weight (g/mol): 242.7
Molecular Formula: C15H11ClO
InChi Key: HIINIOLNGCQCSM-IZZDOVSWSA-N
SMILES: ClC1=CC=C(C=C1)C(=O)\C=C\C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results