missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Chlorochalcone 98.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 97°C |
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 242.70 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4′-Chlorochalcone |
Sécurité et manipulation
| TSCA | No |