accessibility menu, dialog, popup

UserName

CH3H3C(R)OOHN

CAS RN 95530-58-8

IUPAC Name: (4R)-4-(propan-2-yl)-1,3-oxazolidin-2-one
Synonyms: (4R)-4-isopropyl-1,3-oxazolidin-2-one
Molecular Weight (g/mol): 129.16
Molecular Formula: C6H11NO2
InChi Key: YBUPWRYTXGAWJX-YFKPBYRVSA-N
SMILES: CC(C)[C@@H]1COC(=O)N1
Percent Purity 
  • (1)
  • (3)
  • (3)
Quantity 
  • (3)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (1)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)