missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-4-Isopropyl-2-oxazolidinone 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 72°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 129.16 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (R)-4-Isopropyl-2-oxazolidinone |
Safety and Handling
| TSCA | No |