accessibility menu, dialog, popup

UserName

CAS RN 952-92-1

H2NON

CAS RN 952-92-1

IUPAC Name: 1-benzyl-1,4-dihydropyridine-3-carboxamide
Synonyms: 1-benzyl-4H-pyridine-3-carboxamide
Molecular Weight (g/mol): 214.27
Molecular Formula: C13H14N2O
InChi Key: CMNUYDSETOTBDE-UHFFFAOYSA-N
SMILES: NC(=O)C1=CN(CC2=CC=CC=C2)C=CC1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)