missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Benzyl-1,4-dihydronicotinamide 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 121°C |
| Quantity | 5 g |
| Formula Weight | 214.27 |
| Percent Purity | ≥95.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1-Benzyl-1,4-dihydronicotinamide |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |