accessibility menu, dialog, popup

UserName

CAS RN 950761-81-6

IUPAC Name: 4,4,5,5-tetramethyl-2-(perylen-3-yl)-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-(perylen-3-yl)-1,3,2-dioxaborolane
Molecular Weight (g/mol): 378.28
Molecular Formula: C26H23BO2
InChi Key: RKJWQQVQQVALBZ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C2C3=CC=CC4=CC=CC(C5=C2C1=CC=C5)=C34
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results