missing translation for 'onlineSavingsMsg'
Learn More

4,4,5,5-Tetramethyl-2-(3-perylenyl)-1,3,2-dioxaborolane 97.0+%, TCI America™

Catalog No. T3089200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3089200MG 200 mg
T30891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3089200MG Supplier TCI America Supplier No. T3089200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 950761-81-6
Molecular Formula C26H23BO2
Molecular Weight (g/mol) 378.278
InChI Key RKJWQQVQQVALBZ-UHFFFAOYSA-N
Synonym (3-Perylenyl)boronic Acid Pinacol Ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)perylene
PubChem CID 24751681
IUPAC Name 4,4,5,5-tetramethyl-2-perylen-3-yl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C3C=CC=C4C3=C(C=C2)C5=CC=CC6=C5C4=CC=C6

Specifications

Melting Point 237°C
Quantity 200 mg
Formula Weight 378.28
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4,5,5-Tetramethyl-2-(3-perylenyl)-1,3,2-dioxaborolane