accessibility menu, dialog, popup

UserName

CAS RN 95-81-8

IUPAC Name: 2-chloro-5-methylaniline
Synonyms: 2-chlor-5-methyl-aniline
Molecular Weight (g/mol): 141.6
Molecular Formula: C7H8ClN
InChi Key: HPSCXFOQUFPEPE-UHFFFAOYSA-N
SMILES: CC1=CC=C(Cl)C(N)=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)