missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-methylaniline 98.0+%, TCI America™

Catalog No. C25765G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C25765G Supplier TCI America Supplier No. C25765G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 95-81-8
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.60
MDL Number MFCD00007674
InChI Key HPSCXFOQUFPEPE-UHFFFAOYSA-N
Synonym benzenamine, 2-chloro-5-methyl, 6-chloro-m-toluidine, 3-amino-4-chlorotoluene, 2-chlor-5-methyl-aniline, 2-chloro-5-methylbenzenamine, 2-chloro-5-methylphenylamine, ccris 2888, 2-chloro-5-methyl aniline, 6-chlor-meta-toluidin, pubchem18921
PubChem CID 66770
IUPAC Name 2-chloro-5-methylaniline
SMILES CC1=CC=C(Cl)C(N)=C1

Specifications

Color White-Yellow
Boiling Point 230°C
Quantity 5 g
UN Number 2239
Formula Weight 141.60
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Chloro-5-methylaniline
EINECSNumber (3)-0265
TSCA No