accessibility menu, dialog, popup

UserName

CH3ClCl

CAS RN 95-73-8

IUPAC Name: 2,4-dichloro-1-methylbenzene
Synonyms: 2,4-dichlorotoluene
Molecular Weight (g/mol): 161.03
Molecular Formula: C7H6Cl2
InChi Key: FUNUTBJJKQIVSY-UHFFFAOYSA-N
SMILES: CC1=CC=C(Cl)C=C1Cl
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)