missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorotoluene, 99%

Catalog No. AC163282500
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC163282500 250 mL Glass bottle
AC163280010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC163282500 Supplier Thermo Scientific Chemicals Supplier No. 163282500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 95-73-8 | C7H6Cl2 | 161.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 95-73-8
Molecular Formula C7H6Cl2
Molecular Weight (g/mol) 161.03
MDL Number MFCD00000583
InChI Key FUNUTBJJKQIVSY-UHFFFAOYSA-N
Synonym 2,4-dichlorotoluene, benzene, 2,4-dichloro-1-methyl, toluene, 2,4-dichloro, 2,4-dichloro-1-methyl-benzene, unii-3s32n6lg3x, 2,4-dichloromethylbenzene, 2,4-dct, 2,4-diclorotoluene, 4,6-dichlorotoluene, 2,4-dichloro-toluen
PubChem CID 7254
ChEBI CHEBI:81651
IUPAC Name 2,4-dichloro-1-methylbenzene
SMILES CC1=C(C=C(C=C1)Cl)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -14.0°C
Color Colorless
Density 1.2400g/mL
Boiling Point 200.0°C
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5450 to 1.5470
Linear Formula CH3C6H3Cl2
Quantity 250 mL
Beilstein 05, 295
Specific Gravity 1.24
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in alcohol,ether and acetone
Formula Weight 161.03
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 4-Dichlorotoluene, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 202-445-8
RTECSNumber XT0730000
TSCA TSCA

RUO – Research Use Only