accessibility menu, dialog, popup

UserName

OHCl

CAS RN 95-57-8

IUPAC Name: 2-chlorophenol
Synonyms: 2-chlorophenol caswell no. 203 epa pesticide chemical code 062204 septi-kleen ccris 640 rcra waste number U048 pine-O disinfectant
Molecular Weight (g/mol): 128.56
Molecular Formula: C6H5ClO
InChi Key: ISPYQTSUDJAMAB-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1Cl
Boiling Point 
  • (4)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (4)
Percent Purity 
  • (2)
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (7)
  • (4)
  • (2)
  • (3)
  • (5)
  • (4)
  • (2)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)