accessibility menu, dialog, popup

UserName

CAS RN 95-56-7

IUPAC Name: 2-bromophenol
Synonyms: 2-bromophenol bromophenol 2-bromfenol
Molecular Weight (g/mol): 173.01
Molecular Formula: C6H5BrO
InChi Key: VADKRMSMGWJZCF-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1Br
Boiling Point 
  • (3)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (6)
  • (2)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (6)
  • (2)
  • (3)
  • (5)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (5)