missing translation for 'onlineSavingsMsg'
Learn More

2-Bromophenol, 98%

Catalog No. AAA1411236
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1411236 500 g
AAA1411214 25 g
AAA1411222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1411236 Supplier Thermo Scientific Chemicals Supplier No. A1411236
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 95-56-7 | C6H5BrO | 173.009 g/mol

2-Bromophenol used as a disinfection byproduct found in chlorinated pool water. Used in the preparation of anti-benzofurobenzofuran diimides. It was also used to study the photodegradation of 2-bromophenol using UV-Vis spectroscopy and HPLC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromophenol used as a disinfection byproduct found in chlorinated pool water. Used in the preparation of anti-benzofurobenzofuran diimides. It was also used to study the photodegradation of 2-bromophenol using UV-Vis spectroscopy and HPLC.

Solubility
Soluble in chloroform and ether. Slightly soluble in water.

Notes
Light sensitive. Store in dark. Store in cool, dry place in tightly closed containers.

Chemical Identifiers

CAS 95-56-7
Molecular Formula C6H5BrO
Molecular Weight (g/mol) 173.009
MDL Number MFCD00002146
InChI Key VADKRMSMGWJZCF-UHFFFAOYSA-N
Synonym o-bromophenol, phenol, 2-bromo, bromophenol, 2-bromfenol, 2-bromo phenol, 2-bromo-phenol, phenol, o-bromo, o-bromo-phenol, phenol, bromo, 2-bromfenol czech
PubChem CID 7244
IUPAC Name 2-bromophenol
SMILES C1=CC=C(C(=C1)O)Br

Specifications

Melting Point 5°C
Density 1.636
Boiling Point 194°C to 195°C
Flash Point 92°C (192°F)
Odor Phenol-like
Refractive Index 1.589
Quantity 500 g
UN Number UN3082
Beilstein 1905115
Sensitivity Light sensitive
Merck Index 14,1428
Solubility Information Soluble in chloroform and ether. Slightly soluble in water.
Formula Weight 173.01
Percent Purity 98%
Chemical Name or Material 2-Bromophenol
Show More Show Less
Hazard Category H226-H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H315-H319-H335-H227
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 202-432-7
RTECSNumber SJ7875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only