accessibility menu, dialog, popup

UserName

CAS RN 95-33-0

NHSSN

CAS RN 95-33-0

IUPAC Name: N-(1,3-benzothiazol-2-ylsulfanyl)cyclohexanamine
Synonyms: N-(1,3-benzothiazol-2-ylsulfanyl)cyclohexanamine
Molecular Weight (g/mol): 264.41
Molecular Formula: C13H16N2S2
InChi Key: DEQZTKGFXNUBJL-UHFFFAOYSA-N
SMILES: C1CCC(CC1)NSC1=NC2=CC=CC=C2S1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results