missing translation for 'onlineSavingsMsg'
Learn More

N-Cyclohexyl-2-benzothiazolylsulfenamide 98.0+%, TCI America™

Catalog No. C0576500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0576500G 500 g
C057625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0576500G Supplier TCI America Supplier No. C0576500G

Chemical Identifiers

CAS 95-33-0
Molecular Formula C13H16N2S2
Molecular Weight (g/mol) 264.41
MDL Number MFCD00022872
InChI Key DEQZTKGFXNUBJL-UHFFFAOYSA-N
PubChem CID 7232
IUPAC Name N-(1,3-benzothiazol-2-ylsulfanyl)cyclohexanamine
SMILES C1CCC(CC1)NSC1=NC2=CC=CC=C2S1

Specifications

Melting Point 101°C
Quantity 500 g
UN Number 3077
Formula Weight 264.41
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Cyclohexyl-2-benzothiazolylsulfenamide
EINECSNumber (5)-0256
RTECSNumber DL6250000
TSCA Yes