accessibility menu, dialog, popup

UserName

CAS RN 95-20-5

IUPAC Name: 2-methyl-1H-indole
Synonyms: 2-methylindole
Molecular Weight (g/mol): 131.18
Molecular Formula: C9H9N
InChi Key: BHNHHSOHWZKFOX-UHFFFAOYSA-N
SMILES: CC1=CC2=CC=CC=C2N1
Boiling Point 
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)