accessibility menu, dialog, popup

UserName

CAS RN 95-00-1

IUPAC Name: 1-(2,4-dichlorophenyl)methanamine
Synonyms: 1-(2,4-dichlorophenyl)methanamine
Molecular Weight (g/mol): 176.04
Molecular Formula: C7H7Cl2N
InChi Key: SJUKJZSTBBSGHF-UHFFFAOYSA-N
SMILES: NCC1=CC=C(Cl)C=C1Cl
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (4)