missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzylamine 98.0+%, TCI America™

Catalog No. D17405G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D17405G 5 g
D174025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D17405G Supplier TCI America Supplier No. D17405G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 95-00-1
Molecular Formula C7H7Cl2N
Molecular Weight (g/mol) 176.04
MDL Number MFCD00008109
InChI Key SJUKJZSTBBSGHF-UHFFFAOYSA-N
Synonym 2,4-dichlorobenzylamine, 2,4-dichlorophenyl methanamine, benzenemethanamine, 2,4-dichloro, benzylamine, 2,4-dichloro, 2,4-dichlorobenzyl amine, 1-2,4-dichlorophenyl methanamine, 2,4-dichloro-benzylamine, 2,4-dichlorophenyl methylamine, o,p-dichlorobenzylamine, acmc-209rur
PubChem CID 1485
IUPAC Name 1-(2,4-dichlorophenyl)methanamine
SMILES NCC1=CC=C(Cl)C=C1Cl

Specifications

Color Yellow
Quantity 5 g
UN Number 1760
Formula Weight 176.04
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 2,4-Dichlorobenzylamine