accessibility menu, dialog, popup

UserName

HN

CAS RN 948-65-2

IUPAC Name: 2-phenyl-1H-indole
Synonyms: 1H-indole, 2-phenyl-
Molecular Weight (g/mol): 193.25
Molecular Formula: C14H11N
InChi Key: KLLLJCACIRKBDT-UHFFFAOYSA-N
SMILES: N1C2=CC=CC=C2C=C1C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)