accessibility menu, dialog, popup

UserName

NBrBr

CAS RN 94446-97-6

IUPAC Name: 2-bromo-3-(bromomethyl)pyridine
Synonyms: 2-bromo-3-(bromomethyl)pyridine
Molecular Weight (g/mol): 250.92
Molecular Formula: C6H5Br2N
InChi Key: NEWLOSCWMVKPGI-UHFFFAOYSA-N
SMILES: BrCC1=C(Br)N=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results