missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-(bromomethyl)pyridine, 96%, Thermo Scientific Chemicals

Catalog No. AAH52311MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52311MD 250 mg
AAH5231103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52311MD Supplier Thermo Scientific Chemicals Supplier No. H52311MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 94446-97-6
Molecular Formula C6H5Br2N
Molecular Weight (g/mol) 250.921
MDL Number MFCD08276299
InChI Key NEWLOSCWMVKPGI-UHFFFAOYSA-N
Synonym 2-bromo-3-bromomethyl pyridine, 2-bromo-3-bromomethylene pyridine, pubchem22284, 2-bromo-3-bromomethyl-pyridine, pyridine, 2-bromo-3-bromomethyl
PubChem CID 10901078
IUPAC Name 2-bromo-3-(bromomethyl)pyridine
SMILES C1=CC(=C(N=C1)Br)CBr

Specifications

Melting Point 38°C
Boiling Point 291°C
Flash Point >110°C (230°F)
Quantity 250 mg
Formula Weight 250.92
Percent Purity 96%
Chemical Name or Material 2-Bromo-3-(bromomethyl)pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only