accessibility menu, dialog, popup

UserName

CAS RN 943757-71-9

IUPAC Name: 2-{diphenyl[(trimethylsilyl)oxy]methyl}pyrrolidine
Synonyms: 2-{diphenyl[(trimethylsilyl)oxy]methyl}pyrrolidine
Molecular Weight (g/mol): 325.53
Molecular Formula: C20H27NOSi
InChi Key: RSUHWMSTWSSNOW-UHFFFAOYNA-N
SMILES: C[Si](C)(C)OC(C1CCCN1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results