accessibility menu, dialog, popup

UserName

CAS RN 943-80-6

IUPAC Name: (2S)-2-amino-3-(4-aminophenyl)propanoic acid
Synonyms: p-aminophenylalanine
Molecular Weight (g/mol): 180.21
Molecular Formula: C9H12N2O2
InChi Key: CMUHFUGDYMFHEI-QMMMGPOBSA-N
SMILES: N[C@@H](CC1=CC=C(N)C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)