missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-L-phenylalanine, 95%

Catalog No. AAH5197006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5197006 5 g
AAH5197003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5197006 Supplier Thermo Scientific Chemicals Supplier No. H5197006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 943-80-6 | C9H12N2O2 | 180.21 g/mol

It finds its application in the purification of peptide synthetases involved in pristinamycin I biosynthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the purification of peptide synthetases involved in pristinamycin I biosynthesis.

Solubility
Soluble in water.

Notes
Air sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Chemical Identifiers

CAS 943-80-6
Molecular Formula C9H12N2O2
Molecular Weight (g/mol) 180.21
MDL Number MFCD00069927
InChI Key CMUHFUGDYMFHEI-QMMMGPOBSA-N
Synonym 4-amino-l-phenylalanine, h-phe 4-nh2-oh, 4-aminophenylalanine, l-4-aminophenylalanine, p-amino-l-phenylalanine, p-aminophenylalanine, s-2-amino-3-4-aminophenyl propanoic acid, 4-amino-phenylalanine, 2s-2-amino-3-4-aminophenyl propanoic acid, l-4-nh2-phe-oh
PubChem CID 151001
ChEBI CHEBI:29737
IUPAC Name (2S)-2-amino-3-(4-aminophenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(N)C=C1)C(O)=O

Specifications

Solubility Information Soluble in water.
Quantity 5 g
Formula Weight 180.2
Sensitivity Air Sensitive
Percent Purity 95%
Chemical Name or Material 4-Amino-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only